VWR®, Digital Timer, One Channel
Supplier: VWR Collection
VWRI609-0185EA 26.5 SGD
VWRI609-0185
VWR®, Digital Timer, One Channel
Timers Digital Timers
Counts down from 99 minutes, 59 seconds.
  • Loud alarm for noisy environments
  • Memory function
  • Small, portable unit
  • Automatic off function
Delivery information: Supplied with magnet, stand, clip and certificate of calibration.