VWR®, Digital Timer, One Channel
Supplier: VWR Collection
VWRI609-0190EA 48.8 SGD
VWRI609-0190
VWR®, Digital Timer, One Channel
Timers Digital Timers
This single-channel timer is switchable between 19 hours, 59 minutes and 99 minutes, 59 seconds, and has a 3-key operation for ease of use.
  • Count direction: Up/down
  • Last time set memory
  • Audible alarm; displays elapsed time after countdown
Certifications: Certificate of calibration and individual serial number, these electronic timers are tested and calibrated in an exclusive triple-accredited/-registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Delivery information: More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable