VWR®, Digital Timer, Four Channels, with White Board
Supplier: VWR Collection
VWRI609-0208EA 103 SGD
VWRI609-0208
VWR®, Digital Timer, Four Channels, with White Board
Timers Digital Timers
This timer is built in a white board with marker and eraser, which helps to keep track of time and other critical information. It provides different mounting options: flat on bench, vertical view, and metal surface attachment.
  • Switchable count up and down with elapsed time displayed after countdown
  • Switchable capacity of 23 hours, 59 minutes to 59 minutes, 59 seconds
  • Audible alarm for each channel
  • Last time set memory
  • Audible confirmation for a data entry key press
Certifications: Individually tested and inspected using NIST in A2LA accreditedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable