VWR®, Digital Timer, One Channel
Supplier: VWR Collection
VWRI609-0210EA 39.4 SGD
VWRI609-0210
VWR®, Digital Timer, One Channel
Timers Digital Timers
Counts down from 99 minutes, 59 seconds in second increments.
  • Large digits (19 mm) are readable from a distance of 6 metres
  • Three large keys makes it easy to use
  • Alarm sounds for 30 seconds and then silences automatically
  • Memory resets display to last programmed time for quick repeat of frequent, routine tests
Certifications: Compliance with ISO/IEC 17025; accredited by A2LA indicating traceability to standards provided by NIST.
Delivery information: Supplied with magnet, flip-out stand, clip, battery, and certificate.