VWR®, Digital Stopwatch
Supplier: VWR Collection
VWRI609-0212EA 149 SGD
VWRI609-0212
VWR®, Digital Stopwatch
Timers Digital Stopwatches
Quartz-crystal stopwatch with large LCD digits.
  • Timing capacity: 9 hours, 59 minutes, 59 seconds and 99 hundredths
  • Functions: Single action, time out, cumulative split, interval split, continuous roll-over timing and countdown alarm timing
  • Tough, water resistant ABS plastic casing seals out moisture, dust and fumes
  • Ideal for enzyme tests, viscosity measurements, animal studies, and all timed lab tests
Certifications: Compliance with ISO/IEC 17025; accredited by A2LA indicating traceability to standards provided by NIST.
Delivery information: More Product Information