VWR®, Digital Timer, One Channel
Supplier: VWR Collection
VWRI609-0224EA 33.1 SGD
VWRI609-0224
VWR®, Digital Timer, One Channel
Timers Digital Timers
This single-channel timer can be programmed in countdown or count up mode.
  • Timing capacity: 99 minutes, 59 seconds
  • Resolution is 1 second
  • Alarm and memory function
  • Big digit display
Delivery information: Supplied with magnet, clip and stand.
Order Now
SPECIFICATIONS
Environmentally Preferable