VWR® Blue-Steel, Digital Timer, One Channel
Supplier: VWR Collection
VWRI609-0225EA 41.8 SGD
VWRI609-0225
VWR® Blue-Steel, Digital Timer, One Channel
Timers Digital Timers
This timer features a modern design, polished metal case and rubberised backing.
  • Timing capacity: 99 minutes 59 seconds
  • Functions: Count up or count down
  • Automatic Bounceback™ memory for repetitive timing: Even when turned off the timer will retain the programmed timing interval
  • Memory returns to previously programmed setting at touch of a key
  • Alarm: 60 second beep duration
  • Auto shut-off after 2 minutes of inactivity
  • Large sized keys and large display make it easy to set time and read from across the lab
Certifications: A serial numbered certificateMore Product Information