VWR®, Digital Timer, Four Channels
Supplier: VWR Collection
VWRI609-0229EA 65 SGD
VWRI609-0229
VWR®, Digital Timer, Four Channels
Timers Digital Timers
This user-friendly and innovative 100-hour timer fulfills virtually every lab timing requirement. It has four countdown channels, a stopwatch channel, and a time-of-day channel that work with an uncompromising 0,01% accuracy.
  • Timing capacity: Counts up and down for 99 hours, 59 minutes, 59 seconds
  • Unique thru-zero feature allows counting down, alarming, and counting up
  • A time-out mode stops the timer for off periods in both up and down timing
  • T1 through T4 may be started simultaneously or individually
  • Extra-large, ¹/₃" high display and vivid LCD digits
More Product Information