VWR® Giant-Digits™, Digital Countdown Timer
Supplier: VWR Collection
VWRI609-0231EA 55.9 SGD
VWRI609-0231
VWR® Giant-Digits™, Digital Countdown Timer
Timers Digital Timers
View giant size digits on this bench timer from across the lab. Visible from shelves, in hoods, or perched on instruments. Eliminates 'squinting' to observe alarm time remaining.
  • Giant-Digits™ with 33,8 mm digit height; viewable from 50 feet
  • Countdown and alarms on single channel for ease-of-use
  • Three-button intuitive operation eliminates reading instructions
  • Automatic Bounceback™ Memory for repetitive timing
  • Counts up during alarming indicating elapsed time since alarming
  • Times up to 99 minutes, 59 seconds
  • Settable to one second with one-second resolution
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable