VWR®, Disposable Weighing and Drying Dishes or Pans
Supplier: VWR Collection
VWRI611-1350EA 68.8 SGD
VWRI611-1350 VWRI611-1351 VWRI611-1352 VWRI611-1353 VWRI611-1354_B VWRI611-1380 VWRI611-3658 VWRI611-3659 VWRI611-1354
VWR®, Disposable Weighing and Drying Dishes or Pans
Boats Weighing Boats
Dishes and pans, made of aluminium.
  • Compatible with most balances and dryers
  • Oil-free
  • Slightly tapered for easy stacking
Sturdy, disposable dishes and pans. Ideal for general purpose weighing, dispensing, storage and drying applications.