VWR® Density Kits for VWR Balances
Supplier: VWR Collection
VWRI611-2551EA 593 SGD
VWRI611-2551 VWRI611-2553
VWR® Density Kits for VWR Balances
Balances and Scales
Complete kits that, together with the appropriate balance, allow the user to calculate the density of liquids (such as beverages) or non absorbent solids (rocks, polymers, gems, glass, metals etc). The density wizard function is already included as a built-in function of the balance (except for SE models: 611-2605, 611-2606, 611-2607, 611-3236, 611-3246, 611-3299, 611-3344). Density kits are not designed for use with moisture analysers. Both kits contain similar items, the difference is the balance cone attachment:
  • Measuring glass cup 150 mm high, Ø 50 mm, 250 ml
  • Measuring glass cup 95
More Product Information