VWR® Science Education Precision Balances
Supplier: VWR Collection
VWRI611-3236EA 1010 SGD
VWRI611-3236 VWRI611-3260 VWRI611-3299 VWRI611-3468 VWRI611-3476 VWRI611-2602 VWRI611-2604 VWRI611-2607 VWRI611-3860
VWR® Science Education Precision Balances
Balances and Scales
Competitively priced electronic balances for the education sector. All models have a stainless steel top pan.
  • Easy to use
  • Small footprint
  • Selectable measuring units
  • Mains supply or optional external rechargeable battery for portable use
Additional benefits of the models (except 611-2605, 611-2606, 611-2607, 611-3236, 611-3246, 611-3299 and 611-3344) include:
RS232 serial interface
Hook for under-balance weighing suitable for external calibration models only (not for model number ending in i)
Capable of liquid and solid density determination, check-More Product Information