VWR® LAG Advanced, Analytical Balances with Graphic Display
Supplier: VWR Collection
VWRI611-3267EA 2730 SGD
VWRI611-3267 VWRI611-3268 VWRI611-3297 VWRI611-3376 VWRI611-3457 VWRI611-4302
VWR® LAG Advanced, Analytical Balances with Graphic Display
Balances and Scales
An economical range of reliable, high performance analytical balances. All models have a stainless steel top pan.
  • Check weighing (alarm function), weight totalising, density determinations of samples and piece counting functions
  • Models have large graphic display with added functionality for formulation, GLP, date and time, multi-language
  • Option of external or internal calibration, RS232 included as standard
  • Weighing chamber L×W×H: 162×171×225 mm
User can choose three options for models with internal calibration (denoted by 'i' at end of the modelMore Product Information