VWR® Classic LP / LPC / LPW, Precision Balances with LCD
Supplier: VWR Collection
VWRI611-2610EA 1370 SGD
VWRI611-2610 VWRI611-2301 VWRI611-2300 VWRI611-2298 VWRI611-2302 VWRI611-2296 VWRI611-2289 VWRI611-2308 VWRI611-2279 VWRI611-2320 VWRI611-2272 VWRI611-2278 VWRI611-2275 VWRI611-2274 VWRI611-2321 VWRI611-2319 VWRI611-2629 VWRI611-3269 VWRI611-3270 VWRI611-3271 VWRI611-3272 VWRI611-3277 VWRI611-3296 VWRI611-3298 VWRI611-3327 VWRI611-3379 VWRI611-3346 VWRI611-3347 VWRI611-3348 VWRI611-3354 VWRI611-3355 VWRI611-3359 VWRI611-3360 VWRI611-3361 VWRI611-3367 VWRI611-3371 VWRI611-3373 VWRI611-3374 VWRI611-3375 VWRI611-3383 VWRI611-3395 VWRI611-3397 VWRI611-3398 VWRI611-3458 VWRI611-3472 VWRI611-2600 VWRI611-3816 VWRI611-3823 VWRI611-3825 VWRI611-4316 VWRI611-4317 VWRI611-4330 VWRI611-5616 VWRI611-5617
VWR® Classic LP / LPC / LPW, Precision Balances with LCD
Balances and Scales
Rugged and economical balances, that are ideal for educational establishments and laboratory use. A series that ranges from simple weighing to operation with automated internal calibration. All Classic models have a simple backlit LCD with adjustable contrast and ergonomic keypad and a stainless steel top pan.
  • Selectable filter levels and user can select the measuring unit that the balance will use to display the weight
  • RS232 data interface
  • LPW models have a three door draft shield
Functions include suspended weighing, piece counting, percentageMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable