VWR® Professional TP / TPW, Precision Balances with Touch Screen
Supplier: VWR Collection
VWRI611-3692EA 2640 SGD
VWRI611-3692 VWRI611-3693 VWRI611-3688 VWRI611-3689 VWRI611-3690 VWRI611-3694 VWRI611-3700 VWRI611-3701 VWRI611-3695 VWRI611-3696 VWRI611-3697 VWRI611-3703 VWRI611-3698 VWRI611-3699 VWRI611-3704 VWRI611-3706 VWRI611-3826 VWRI611-3827 VWRI611-3858 VWRI611-3857 VWRI611-3861 VWRI611-3862 VWRI611-4677 VWRI611-4678
VWR® Professional TP / TPW, Precision Balances with Touch Screen
Balances and Scales
VWR® Professional precision balances are ideal for wide range of laboratory weighing tasks, they have been designed for multi-user environments (each user profile has password protection). These interactive balances, which feature an internal database to store data and drying methods, help improve efficiency and productivity. All professional models have a 5" touch screen with icon-driven menu, qwerty touch keypad and a stainless steel top pan. Models are GLP set-up, storing balance, project and user identification data, as well as date and time. GLP information can be also printed out withMore Product Information