VWR® TA Professional, Analytical Balances with Touch Screen
Supplier: VWR Collection
VWRI611-3677EA 3850 SGD
VWRI611-3677 VWRI611-3684 VWRI611-3686 VWRI611-3683 VWRI611-3679 VWRI611-3681 VWRI611-3674 VWRI611-3676 VWRI611-3828 VWRI611-3829 VWRI611-3830 VWRI611-3831
VWR® TA Professional, Analytical Balances with Touch Screen
Balances and Scales
Laboratory analytical balances that have 5" colour touch screen display with advanced applications and large databases for professional weighing. The user has easy and intuitive access to balance functions, via interactive, icon-driven menus ensuring efficient and productive weighing. Balances can be used by up to 10 different users, each with their own customisable settings and password protection. Ideal for laboratories requiring GLP compliance. All models have a stainless steel top pan.
  • Internal and external calibration (optional factory calibration
More Product Information