VWR®, Weighing Boats, Sterile
Supplier: VWR Collection
VWRI611-4672EA 236 SGD
VWRI611-4672 VWRI611-5272 VWRI611-5273 VWRI611-5274
VWR®, Weighing Boats, Sterile
Boats Weighing Boats
These disposable boats are flexible and feature smooth surfaces to provide accurate pour-outs with minimal sample loss. Ideal for applications where there is concern of cross-contamination and 'clean' is a necessity in life science research, food, biopharmaceutical, environmental and molecular research environments. Manufactured from FDA food grade PS and packed in a cleanroom.
  • Electron beam sterilised
  • Lot number traceability for quality assurance
  • Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases
More Product Information