VWR® Professional, Pipette Calibration Balances
Supplier: VWR Collection
VWRI611-4674EA 4740 SGD
VWRI611-4674 VWRI611-4673 VWRI611-4675 VWRI611-4676
VWR® Professional, Pipette Calibration Balances
Balances and Scales
Pipetting accuracy is critical for successful experiments: A small inaccuracy in pipetting can result in important variations in results. The pipette check function of these touchscreen balances allows the user to check if the pipette is still accurate, as per directive ISO 8655 'Piston-operated volumetric apparatus - Gravimetric methods for the determination of measurement error' and customisable methods.
  • Balances can be used for standard weighing operations as well as pipette checking
  • Inbuilt software guides the user in selecting which volume to check,
More Product Information