VWR®, Dot Printer for VWR® Balances
Supplier: VWR Collection
VWRI611-4790EA 843 SGD
VWRI611-4790
VWR®, Dot Printer for VWR® Balances
Printers Document Printers
The TX-110 provides high speed dot printing and features a 184×64 OLED display.
  • Printouts are suitable for long-term preservation
  • ISO/GLP compliant data printout
  • Built-in real-time clock
  • Sample name and batch number can be input and printed
  • Sum, mean and formulation function