VWR® Safety Bottle Carriers
Supplier: VWR Collection
VWRI611-5280EA 106 SGD
VWRI611-5280 VWRI611-5281 VWRI611-5282 VWRI611-5283 VWRI611-5284 VWRI611-5285 VWRI611-5286 VWRI611-5287
VWR® Safety Bottle Carriers
Carriers Bottle Carriers/Containers
The safety bottle carrier provides a safe and reliable way to transport a variety of bottled reagents in the lab. Available in 1,5 and 4,5 L capacities, this carrier is the practical answer for any clinical and research lab setting.
  • Ergonomic handle prevents hand fatigue and related accidents when carrying heavier loads
  • Handle can be pressed down to either side for easier loading and unloading
  • Centering ring stabilises bottle in the centre of carrier for safer, steadier transport
  • Carriers come in four colours with designated labelling areas for easy
More Product Information