VWR® Anti-Vibration Tables
Supplier: VWR Collection
VWRI611-5596EA 4450 SGD
VWRI611-5596 VWRI611-5597
VWR® Anti-Vibration Tables
Furniture Tables and Workbenches
The anti-vibration table provides a stable and vibration-free surface for sensitive equipment, such as microscopes, balances, or other precision instruments. It helps to minimise external vibrations from affecting the performance and accuracy of the equipment, ensuring reliable measurements and observations.
  • The integrated granite isolation area sits flush with the surrounding work area (standard), maximising available space
  • Provides suitable working conditions for sensitive devices
  • Large working surface of 900×600 mm with integrated granite slab (500×350
More Product Information