VWR®, Pour-Boat Weighing Dishes
Supplier: VWR Collection
VWRI611-9163EA 59.8 SGD
VWRI611-9163 VWRI611-9164 VWRI611-9165 VWRI611-3116 VWRI611-3117 VWRI611-3118
VWR®, Pour-Boat Weighing Dishes
Boats Weighing Boats
PS.
  • Flat bottom avoids tipping
  • Temperature resistant to 70 °C
  • Maximise recovery of often very expensive chemicals
  • Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols and bases
Samples are easily dispensed with the pour spout design without flexing the dish. These flat bottomed dishes are made from 0,3 mm thick polystyrene for better strength, stability and handling.
The anti-static versions help reduce the risk of static build up on samples, and helps maximise sample transfer when weighing out products.