VWR®, Disposable Weighing Boats
Supplier: VWR Collection
10770-450
VWRI611-0093EA 36.5 SGD
VWRI611-0093 VWRI611-0094 VWRI611-0095 VWRI611-9168 VWRI611-9169 VWRI611-9170 VWRI611-9175 VWRI611-9176 VWRI611-9177 VWRI611-9178 VWRI611-9179 VWRI611-9180 VWRI611-9181 VWRI611-9182 VWRI611-9183 VWRI611-3113 VWRI611-3114 VWRI611-3115 VWRU10770-450
VWR®, Disposable Weighing Boats
Boats Weighing Boats
PS, square.
  • Shallow form with wide, flat bottom avoids tipping
  • Rounded corners simplify transfer
  • Temperature resistant up to 70 °C
These flexible weighing boats, with smooth surfaces, enable accurate pour-out with minimal sample loss. They are biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols and bleaches. Ideal for use with microwaves, balances or moisture analysers.
Packaging: These weighing boats are produced and packed in plastic bags in a cleanroom.