VWR®, Disposable Weighing Boats
Supplier: VWR Collection
VWRI611-0097EA 39.2 SGD
VWRI611-0097 VWRI611-9184 VWRI611-9185 VWRI611-9186 VWRI611-9187 VWRI611-9188 VWRI611-9189 VWRI611-9190 VWRI611-9191 VWRI611-9192 VWRI611-9161 VWRI611-9162 VWRI611-1993 VWRI611-1994 VWRI611-1995 VWRI611-1996
VWR®, Disposable Weighing Boats
Boats Weighing Boats
PS, diamond.
  • Wide, flat bottom avoids tipping
  • Temperature resistant up to +70 °C
These flexible boats feature smooth surfaces to provide accurate pour-out with minimal sample loss. They are ideal for use with microwaves and moisture analysers.
Packaging: They are produced and packed in plastic bags in a cleanroom.
Order Now
SPECIFICATIONS
Environmentally Preferable