VWR®, Straw Pipettes
Supplier: VWR Collection
VWRI612-1675EA 88.8 SGD
VWRI612-1675
VWR®, Straw Pipettes
Pipettes Straw Pipettes
Straw pipettes made from PP, irradiated, disposable, sterile, without graduation.
  • Traceability including the expiry date on the secondary packaging
  • Zip bag can be pressed closed ensuring product protection
  • Tamper-evident seal guarantees the sterility of the product
Caution: *The product(s) below are outside the scope of the EU Directive 2019/904