VWR®, Safety Pipette Fillers, Standard/Universal/Flip
Supplier: VWR Collection
53497-055
VWRI612-1930EA 14.8 SGD
VWRI612-1930 VWRI612-2008 VWRI612-1931 VWRI612-2004 VWRI612-1920 VWRI612-1947 VWRU53497-055
VWR®, Safety Pipette Fillers, Standard/Universal/Flip
Pipettes Pipette Bulbs
Natural rubber.
  • For measuring and for volumetric pipettes up to 10 ml (standard) or 100 ml (universal/flip)
  • Standard/universal: Three ventilation valves
  • Flip: Only two operating points for easier usage (release is through an automatic removable valve)