VWR®, Pipette Bulbs
Supplier: VWR Collection
VWRI612-2691EA 60.3 SGD
VWRI612-2691 VWRI612-2693
VWR®, Pipette Bulbs
Pipettes Pipette Bulbs
Natural latex.
  • For Pasteur pipettes, small pipettes and medical drop counters
Capacity: 1 to 2 ml
Length: 43/52 mm