VWR®, Safety Bulb Pipette Fillers
Supplier: VWR Collection
VWRI612-2703EA 36.4 SGD
VWRI612-2703
VWR®, Safety Bulb Pipette Fillers
Pipettes Pipette Bulbs
Red rubber with white plastic connector and silicone conical adapter, autoclavable.
  • For large and small capacity pipettes
  • Conical adapter connects pipettes more securely, avoids breakages because it requires less force to fit on the bulb
  • Elastic cord for attaching the bulb to a reagent vessel or to a user's wrist
Capacity: 15 ml