VWR®, Vacuum/Pressure Bulb
Supplier: VWR Collection
VWRI612-2705EA 29.3 SGD
VWRI612-2705
VWR®, Vacuum/Pressure Bulb
Pipettes Pipette Bulbs
Black rubber with PVC connectors.
  • For collecting gas-phase samples or aspirating gases through instruments
  • Two thick-walled rubber valves
Capacity: 85 ml
Colour: black
Ordering information: Supplied without tubing.
Order Now
SPECIFICATIONS
Environmentally Preferable