VWR® MultiTray®/MultiBox® plus, Pipette Tips
Supplier: VWR Collection
VWRI612-2914EA 85.8 SGD
VWRI612-2914 VWRI612-2915 VWRI612-2599 VWRI612-2913 VWRI612-2999
VWR® MultiTray®/MultiBox® plus, Pipette Tips
Pipette Tips
Tips racked in MultiTrays®. Compatible with most micropipettes, except 612-2913 which is compatible with Gilson® and Labmate® micropipettes. MultiBox® plus is an autoclavable PP box with dustproof, hinged and sliding lid, which may be opened completely or partially. MultiBox® plus is suitable for use with all MultiTrays®.
  • Tips fit firmly on micropipettor cones, have straight shape for optimum handling, and offer minimum fluid retention
  • MultiBox® plus can be repeatedly autoclaved, but should be placed in an adapted container to avoid contact with the
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable