VWR® Powerpette Plus, Pipette Controller
Supplier: VWR Collection
VWRI612-3686EA 412 SGD
VWRI612-3686
VWR® Powerpette Plus, Pipette Controller
Pipettes Pipette Controllers
Pipette controller for glass and plastic pipettes from 1 to 100 ml.
  • Speed of suction and dispensing controlled through the finger triggers
  • Three aspirate and dispense modes
  • High and low settings both have variable speed
  • Fully charged has approximately 8 hours of continuous use available
  • Low battery light illuminates when 1 hour's power remains
Offers a unique combination of features to give precision fingertip control in a classic streamlined design, suitable for users with large or small hands. Gravity dispense mode is designed for use with 'ToMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable