VWR® Volumetric Flasks, Class A, Glass, Amber, NS Plastic Stopper
Supplier: VWR Collection
VWRI612-3826EA 106 SGD
VWRI612-3826 VWRI612-3825 VWRI612-3824 VWRI612-3823 VWRI612-3822 VWRI612-3821 VWRI612-3828 VWRI612-3829 VWRI612-3827 VWRI612-3831 VWRI612-3830
VWR® Volumetric Flasks, Class A, Glass, Amber, NS Plastic Stopper
Flasks Volumetric Flasks
Borosilicate 3.3 glass, amber, class A.
  • White PE stopper
  • Marks and inscriptions in white enamel
  • Calibrated to contain (TC, In)
DIN EN ISO 1042
Ordering information: Batch number and batch certificate included.
Order Now
SPECIFICATIONS
Environmentally Preferable