VWR®, Bulb Pipettes, One Mark, Class AS
Supplier: VWR Collection
VWRI612-4133EA 32.3 SGD
VWRI612-4133 VWRI612-4129 VWRI612-4130 VWRI612-4132 VWRI612-4134 VWRI612-4135 VWRI612-4136 VWRI612-4137 VWRI612-4138 VWRI612-4131
VWR®, Bulb Pipettes, One Mark, Class AS
Pipettes Bulb Pipettes
Borosilicate 9.1 glass.
  • Calibrated to deliver (TD)
  • Ring mark and inscriptions in high contrast amber stain
  • Not available with individual certificate
Certifications: DIN EN ISO 648
Ordering information: Dated batch identification and batch certificate included.