VWR® Graduated Pipettes, Type 3, Class AS
Supplier: VWR Collection
VWRI612-4767EA 16.2 SGD
VWRI612-4767 VWRI612-4768 VWRI612-4769 VWRI612-4770 VWRI612-4771 VWRI612-4772 VWRI612-4773 VWRI612-4774
VWR® Graduated Pipettes, Type 3, Class AS
Pipettes Graduated Pipettes
Borosilicate 9.1 glass.
  • Calibrated to deliver (TD, Ex)
  • Marks and inscriptions in amber stain
  • Cotton plug ending (cotton plugs not included)
Certifications: DIN 12696, ISO 835
Ordering information: Includes a lot certificate.
Order Now
SPECIFICATIONS
Environmentally Preferable