VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
VWRI612-5504EA 78.3 SGD
VWRI612-5504 VWRI612-5505 VWRI612-5506 VWRI612-5507 VWRI612-5521 VWRI612-5522 VWRI612-5523 VWRI612-5539 VWRI612-5540 VWRI612-5541 VWRI612-5542 VWRI612-5543 VWRI612-5544 VWRI612-5545 VWRI612-5546 VWRI612-5547 VWRI612-5548 VWRI612-5826 VWRI612-5827 VWRI612-5828 VWRI612-5833 VWRI612-5830 VWRI612-5834 VWRI612-5835 VWRI612-5831 VWRI612-5836 VWRI612-5829 VWRI612-5837 VWRI612-5832
VWR® Premium Line, Serological Pipettes
Pipettes Serological Pipettes
PS, transparent, graduated, non sterile or sterile. These disposable pipettes for liquid transfers are made of GPPS (general polystyrene), in compliance with USP Class VI.
  • DNase-/RNase-free, non pyrogenic, non cytotoxic, non haemolytic
  • Meet the requirement of BST/TSE; latex-free
  • Graduations are calibrated for accurate dispensing to within ±2%
  • Pipettes are colour coded according to their volume
  • Sterile pipettes: Sterilised by irradiation
  • Printed with a batch number for traceability
Manufactured in a class 100 000 cleanroom environment. ManufacturedMore Product Information