VWR® Pipette Holder, Wall Mountable
Supplier: VWR Collection
VWRI612-5779EA 93.2 SGD
VWRI612-5779 VWRI612-5783
VWR® Pipette Holder, Wall Mountable
Pipette Stands
This universal wall mount made from robust ABS holds two single channel, or two multi-channel pipettes, or a combination of both.
  • Can securely hold virtually every known brand of pipette
  • Inserts on the upper segments can be added or removed to accommodate the fit of various pipette widths
  • Enables one handed removal or replacement of pipettes
  • Pipettes are held in a vertical position to help prevent contamination
  • Unit is not autoclavable but can be cleaned with soap/water or common laboratory disinfectants
The pipette holder comes with two wall mountingMore Product Information