VWR® Burettes, with Detachable PTFE Stopcock
Supplier: VWR Collection
VWRI612-6694EA 93.4 SGD
VWRI612-6694 VWRI612-6695 VWRI612-6696
VWR® Burettes, with Detachable PTFE Stopcock
Burettes
Borosilicate 3.3 glass, class AS.
  • High chemical and thermal resistance
  • Schellbach stripe for accurate meniscus reading
  • Detachable and interchangeable PTFE stopcock
  • High contrast blue enamel graduations
  • Batch certificate included
No breakage in case of improper handling because the stopcock is not directly connected with the body of the burette. Fast to dismantle, easy to clean and simple to repair because all individual components are replaceable.
Certifications: DIN EN ISO 385