VWR®, Measuring Flasks With Single Certificate
Supplier: VWR Collection
VWRI612-6701EA 18.6 SGD
VWRI612-6701 VWRI612-6702 VWRI612-6703 VWRI612-6704 VWRI612-6705 VWRI612-6706 VWRI612-6707 VWRI612-6708 VWRI612-6709 VWRI612-6710 VWRI612-6711
VWR®, Measuring Flasks With Single Certificate
Flasks Volumetric Flasks
Borosilicate 3.3 glass, with PP stopper. Single certified class A measuring flasks with imprinted serial number and individual work certificate.
  • High chemical and thermal resistance
  • Delivered with white PP stopper
  • Inscriptions and ring mark in high contrasting blue enamel
  • Calibrated to contain (TC, In)
  • Imprinted serial number and single certificate included
Made from true bore glass tubing to guarantee exact measurement.
Certifications: DIN EN ISO 1042. Conformity certified with DE-M mark.