VWR® Bulb Pipettes, One Mark, Class AS, with Single Certificate
Supplier: VWR Collection
VWRI612-6712EA 16.7 SGD
VWRI612-6712 VWRI612-6713 VWRI612-6714 VWRI612-6715 VWRI612-6716 VWRI612-6717 VWRI612-6718 VWRI612-6719 VWRI612-6720 VWRI612-6721
VWR® Bulb Pipettes, One Mark, Class AS, with Single Certificate
Pipettes Bulb Pipettes
Borosilicate 9.1 glass. Single certified class AS bulb pipettes with one mark, imprinted serial number and individual work certificate.
  • Calibrated to deliver (TD, Ex)
  • Inscriptions and ring mark in high contrasting amber
  • Imprinted serial number, single certificate included
Made from true bore glass tubing to guarantee exact measurement at each measuring point. Colour coded and conformity certified with DE-M mark.
Certifications: DIN EN ISO 648