VWR® Pipette Filler, White/Blue
Supplier: VWR Collection
VWRI612-7284EA 87.8 SGD
VWRI612-7284
VWR® Pipette Filler, White/Blue
Pipettes Pipette Controllers
The VWR® pipette filler provides precise fluid control at your fingertips.
  • A T-bar thumb lever controls the aspirating and dispensing speeds from serological pipettes from 1 to 100 ml in size
  • Conveniently accommodates either glass or plastic serological or volumetric pipettes ranging from 1 to 100 ml
  • Includes a replaceable 0,45 μm hydrophobic filter to protect your samples and internal components
  • Inline ergonomic design reduces stress with ease of handling and operation
Comfortable to hold and easy to use, the manual inline pipette controller wasMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Body material ABS