VWR®, Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWRI612-9358EA 88.1 SGD
VWRI612-9358
VWR®, Inoculating Loops and Needles, Disposable
Spreaders Inoculating Loops and Needles
PS, sterile. These loops ensure consistent wetting and complete liquid transfer. Available rigid or flexible and have smooth edges to avoid gouging the surface of the media.
  • Gamma sterilised
  • Colour coded
  • Hexagonal shaft for easy handling
Delivery information: Supplied with a calibration certificate in each box.