VWR®, Single Channel Pipettes, Electronic, Variable Volume
Supplier: VWR Collection
VWRI613-5415EA 1340 SGD
VWRI613-5415 VWRI613-5416 VWRI613-5417 VWRI613-5418 VWRI613-5781
VWR®, Single Channel Pipettes, Electronic, Variable Volume
Pipettes Micropipettes
These pipettes combine superior accuracy, precision and robustness with advanced ergonomic design.
  • Ease of use: Only two buttons and five pipetting modes; enables an easy one-handed programming and a quick pipetting cycle start
  • CUSTOM mode: Users can quickly and easily create specific protocols thanks to VWR® Protocol Builder software
  • Advanced ergonomics: Lightweight and perfectly balanced, it requires virtually no force to significantly reduce RSI
  • Fits comfortably in either the left or right hand and allows a 360° rotating manifold
  • Fully motorised piston
More Product Information