VWR® Pipette Stand, Rotary
Supplier: VWR Collection
VWRI613-6077EA 287 SGD
VWRI613-6077 VWRI612-6117
VWR® Pipette Stand, Rotary
Pipette Stands
This carousel pipette stand, made from ABS, can hold up to eight single-channel, four multi-channel, or a combination of both pipettes.
  • Weighted base allows for easy removal or replacement of pipettes with one hand
  • Pipettes are held in a vertical position to help prevent contamination
  • Conveniently rotate stand head to select or store pipettes
  • Quick grip handle to move pipette stand (loaded or unloaded)
  • Unit is not autoclavable, but can be cleaned with soap/water or common laboratory disinfectants
A sleek, modern design enables this stand to securelyMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable