VWR®, LTS® Compatible Pipette Tips
Supplier: VWR Collection
VWRI613-6516EA 92.2 SGD
VWRI613-6516 VWRI613-6495 VWRI613-6496 VWRI613-6497 VWRI613-6498 VWRI613-6499 VWRI613-6502 VWRI613-6503 VWRI613-6504 VWRI613-6505 VWRI613-6506 VWRI613-6507 VWRI613-6508 VWRI613-6509 VWRI613-6510 VWRI613-6511 VWRI613-6512 VWRI613-6513 VWRI613-6514 VWRI613-6515
VWR®, LTS® Compatible Pipette Tips
Pipette Tips
VWR® LTS® compatible pipette tips feature a flexible seal area and a positive stop resulting in a secure seal with light ejection force on LTS® compatible pipettors.
  • Compatible with the LTS® style pipettors
  • Maximises sample recovery with low binding resin and a fine point design
  • Improves ergonomics by reducing the force required to attach and eject a tip
  • Autoclavable at 1 bar, 122 °C for 15 minutes
  • Manufactured under an ISO 9001 quality management system
VWR® tips use a low retention resin that is up to ten times more hydrophobic than otherMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Waste Reducing,Sustainable Packaging
Environmental certification FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic