VWR® LTS Compatible Adjustable Volume Pipettors
Supplier: VWR Collection
VWRI613-6918EA 2760 SGD
VWRI613-6918 VWRI613-6919 VWRI613-6920 VWRI613-6921 VWRI613-6922 VWRI613-6923 VWRI613-6924 VWRI613-6925 VWRI613-6926 VWRI613-6927 VWRI613-6928 VWRI613-6929 VWRI613-6930 VWRI613-6931 VWRI613-6932
VWR® LTS Compatible Adjustable Volume Pipettors
Pipettes Micropipettes
The VWR® LTS compatible pipettors reduce ejection force with a unique positive stop technology. This technology along with lighter plunger springs, a durable light weight design, and a comfortable handle design with perfect symmetry makes this the best choice for a lab trying to improve ergonomics.
  • Fully compatible with Rainin LTS tips
  • Lighter springs give a more comfortable and controlled draw and dispense
  • Constructed with durable lightweight materials
  • The ejector is cushioned to reduce dynamic stress on your thumb
  • Precision piston design produces
More Product Information