VWR®, Macro Pipette Tips
Supplier: VWR Collection
VWRI613-7094EA 49.2 SGD
VWRI613-7094 VWRI732-3912 VWRI613-7095 VWRI732-3913 VWRI613-7096 VWRI732-3914 VWRI613-7097 VWRI732-3915 VWRI613-7098 VWRI613-7099 VWRI613-7100 VWRI732-3916 VWRI613-7101 VWRI732-3917
VWR®, Macro Pipette Tips
Pipette Tips
Improved accuracy over serological pipettes.
  • Tip length facilitates access into bottles, cell culture flasks, and tubes
Universal fit for compatibility with most major pipettor brands.
Certifications: Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free. Sterile product is certified pyrogen-free.
Order Now
SPECIFICATIONS
Environmentally Preferable