VWR® Standard Line, Multi-Channel Pipettes, Mechanical, Variable Volume
Supplier: VWR Collection
VWRI613-7111EA 676 SGD
VWRI613-7111 VWRI613-7112 VWRI613-7113 VWRI613-7114 VWRI613-7115 VWRI613-7116 VWRI613-7117 VWRI613-7118
VWR® Standard Line, Multi-Channel Pipettes, Mechanical, Variable Volume
Pipettes Micropipettes
These lightweight Standard Line pipettes feature a revolutionary suspension system which allows the shaft to retract slightly when pressed against a row of pipette tips. Each shaft moves independently, resulting in an even sealing force being applied to all 8 or 12 tips. The suspension system not only ensures secure tip sealing, but also eliminates tips being jammed.
  • Easily adjustable ejector height system to accommodate virtually all brands of tips
  • Contoured shape of the handle, 360° manifold and soft spring system delivers smooth and effortless pipetting,
More Product Information