VWR® Reagent Reservoirs
Supplier: VWR Collection
VWRI613-7284EA 53.9 SGD
VWRI613-7284 VWRI613-7285 VWRI613-7286 VWRI613-7287 VWRI613-7289 VWRI613-7290 VWRI613-7291 VWRI613-7293 VWRI613-7294 VWRI613-7297 VWRI613-7299 VWRI613-7301 VWRI613-7304 VWRI613-7305 VWRI613-7306 VWRI613-7307 VWRI613-7308 VWRI613-7310 VWRI613-7311 VWRI613-7312 VWRI613-7314
VWR® Reagent Reservoirs
Reagent Reservoirs
Reagent reservoirs are designed for efficient liquid transfer, making them an ideal choice for repetitive pipetting tasks. These reagent reservoirs for multichannel pipettes provide a trough wide enough to fit 8 and 12 channel pipettes.
  • Trough design with graduations on the inside wall and pour-off spouts on four corners
  • Slightly tilted inner surface helps reduce residue
  • Non sterile or sterilised options available, achieving SAL 10⁻⁶ (ISO 11137)
  • Reservoirs are lot-certified, non-pyrogenic, DNase- and RNase-free
VWR® reagent reservoirs are an excellentMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M