VWR®, Environmentally Friendly Thermometers, Easy Read
Supplier: VWR Collection
VWRI620-0781EA 31.6 SGD
VWRI620-0781 VWRI620-0782 VWRI620-0784 VWRI620-0785 VWRI620-0786 VWRI620-0787 VWRI620-0788
VWR®, Environmentally Friendly Thermometers, Easy Read
Thermometers Analogue Thermometers
These thermometers are easy to read quickly and accurately due to the high contrast black numbering against a yellow background. Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
  • Enviro safe liquid
  • Eliminates reading errors
  • Non toxic, non hazardous, biodegradable liquid
Traceable to NIST/DKD. Compliant with Envirokleen™.
Certifications: Traceable NIST. Envirokleen certified by ChemTel (an independentMore Product Information