VWR®, Frio Temp Precision Thermometers
Supplier: VWR Collection
VWRI620-0843EA 169 SGD
VWRI620-0843 VWRI620-0844 VWRI620-0845 VWRI620-0848 VWRI620-0854 VWRI620-0855 VWRI620-0856 VWRI620-0878 VWRI620-0886 VWRI620-0902
VWR®, Frio Temp Precision Thermometers
Thermometers Analogue Thermometers
These glass thermometers can be used to take readings in freezers, refrigerators, incubators, ovens and blood banks.
  • Enviro safe non toxic and liquid or red spirit liquid
  • Accurate temperature reading without sample contamination
  • Individual serial number
  • Wide choice
Accurate temperature reading is achieved by using the filled bottle (included), which mimics the sample. Insert the thermometer into the bottle and place near the samples for best results. PFA coating safely contains glass and liquid in case of breakage.
Traceable to NIST/DKD
Order Now
SPECIFICATIONS
Environmentally Preferable Safer Human and Environmental Health